Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serpin A6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4065
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4065 100 μg
MAB4065SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4065 Supplier R&D Systems Supplier No. MAB4065
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Serpin A6/Cortisol Binding Globulin Monoclonal specifically detects Serpin A6/Cortisol Binding Globulin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Serpin A6/Cortisol Binding Globulin
Applications Western Blot
Classification Monoclonal
Clone 433903
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q06770
Gene Alias antitrypsin), member 6, CBG, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin
Gene Symbols SERPINA6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin A6 Val23-Ala397 Accession # Q06770
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Test Specificity Detects mouse Serpin A6 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Serpin A1, A5, B8, C1, D1, E1, E2, F1, F2, I1, recombinant human Serpin A3, A4, B6, B9, G1, I2, or recombinant rat Serpin A6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.