Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serpin A8/Angiotensinogen Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6966
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6966 Supplier R&D Systems Supplier No. MAB6966
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Serpin A8/Angiotensinogen Monoclonal specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 754438
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11859
Gene Alias AGT, alpha-1 antiproteinase, antitrypsin, angiotensin II, Angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, Aogen, FLJ92595, FLJ97926, PAT, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Gene Symbols AGT
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Serpin A8/Angiotensinogen Asp25-Val477 Accession # P11859
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 183
Test Specificity Detects mouse Serpin A8/Angiotensinogen in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A8/Angiotensinogen, rhSerpin A4, recombinant mouse (rm) Serpin A1, A3N, A11, B8, C1, E1, E2, I1, or rmPEDF is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.