Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serpin C1/Antithrombin-III Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1287SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1287SP 25 μg
MAB1287 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1287SP Supplier R&D Systems Supplier No. MAB1287SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Serpin C1/Antithrombin-III
Applications Western Blot
Classification Monoclonal
Clone 208730
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P32261
Gene Alias Antithrombin-III, AT3antithrombin-III, ATIII, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Gene Symbols SERPINC1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin C1/Antithrombin-III His33-Asn465 Accession # P32261
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 462
Test Specificity Detects mouse Serpin C1/Antithrombin-III in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human (rh) Serpin C1 is observed and no cross-reactivity with rhSerpin A1, A3, F2, recombinant mouse Serpin D1 or A5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.