Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serpin F2/alpha 2-Antiplasmin Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1239
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1239 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1239 Supplier R&D Systems Supplier No. BAF1239
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin F2/alpha 2-Antiplasmin Polyclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Serpin F2/alpha 2-Antiplasmin
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q61247
Gene Alias AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Gene Symbols SERPINF2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin F2/alpha 2-Antiplasmin Val28-Lys491 Accession # Q61247
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5345
Test Specificity Detects mouse Serpin F2/alpha 2-Antiplasmin in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Serpin F2 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Serpin C1, rmSerpin D1, rhSerpin A1, and rhSerpin C1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.