Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serpin I1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5054
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5054 100 μg
AF5054SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF5054 Supplier R&D Systems Supplier No. AF5054
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin I1/Neuroserpin Polyclonal specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Serpin I1/Neuroserpin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Alias DKFZp781N13156, Neuroserpin, Peptidase inhibitor 12, PI12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1
Gene Symbols SERPINI1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin I1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5274
Test Specificity Detects mouse Serpin I1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Serpin I1 is observed and less than 5% cross-reactivity with rhSerpin I2is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.