Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serum Amyloid A1/A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2948SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2948SP 25 μg
MAB2948100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2948SP Supplier R&D Systems Supplier No. MAB2948SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen SerumAmyloidA1/A2
Applications ELISA
Classification Monoclonal
Clone 834835R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P05366
Host Species Rat
Immunogen E. coli-derived recombinant mouse Serum Amyloid A1/A2 Gly20-Tyr122 Accession # P05366
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 6288
Test Specificity Detects mouse Serum Amyloid A1/A2 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.