Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SEZ6L Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4804SP
This item is not returnable.
View return policy
Description
SEZ6L Polyclonal specifically detects SEZ6L in Mouse samples. It is validated for Immunohistochemistry.Specifications
SEZ6L | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Acig1, AIG1, Aig1L, BSRP-B, seizure related 6 homolog (mouse)-like, seizure related gene 6-like, SEZ6L | |
Sheep | |
Affinity Purified | |
RUO | |
23544 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
AAH65117 | |
SEZ6L | |
Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L Glu32-Ser889 Accession # AAH65117 | |
25 μg | |
Primary | |
Detects mouse SEZ6L in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) BSRP-A and rmBSRP-C is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction