Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SEZ6L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4804
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4804 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4804 Supplier R&D Systems Supplier No. AF4804
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SEZ6L Polyclonal specifically detects SEZ6L in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen SEZ6L
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. AAH65117
Gene Alias Acig1, AIG1, Aig1L, BSRP-B, seizure related 6 homolog (mouse)-like, seizure related gene 6-like, SEZ6L
Gene Symbols SEZ6L
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L Glu32-Ser889 Accession # AAH65117
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23544
Test Specificity Detects mouse SEZ6L in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) BSRP-A and rmBSRP-C is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.