Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SEZ6L2/BSRP-A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4916SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4916SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4916SP Supplier R&D Systems Supplier No. MAB4916SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

SEZ6L2/BSRP-A Monoclonal specifically detects SEZ6L2/BSRP-A in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SEZ6L2/BSRP-A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 529805
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q4V9Z5
Gene Alias BSRPA, BSRP-A, PSK-1, seizure 6-like protein 2, seizure related 6 homolog (mouse)-like 2, UNQ1903/PRO4349
Gene Symbols SEZ6L2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L2/BSRP-A Leu28-Ser837 Accession # Q4V9Z5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26470
Test Specificity Detects mouse SEZ6L2/BSRP-A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) BSRP-B or rmBSRP-C is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.