Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Shrew-1/AJAP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7970
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7970 Supplier R&D Systems Supplier No. AF7970
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 3 publications

Shrew-1/AJAP1 Polyclonal specifically detects Shrew-1/AJAP1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Shrew-1/AJAP1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. A2ALI5
Gene Alias adherens junction associated protein 1, adherens junction-associated protein 1, adherens junctions associated protein 1, AJAP1, Gm573, membrane protein shrew-1, MOT8, RP3-426F10.1, Shrew1, SHREW-1, transmembrane protein SHREW1
Gene Symbols AJAP1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Shrew-1/AJAP1 His157-His283 Accession # A2ALI5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55966
Test Specificity Detects mouse Shrew-1/AJAP1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human AJAP1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.