Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Siglec-1/CD169 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5610T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5610T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5610T100 Supplier R&D Systems Supplier No. FAB5610T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-1/CD169
Applications Flow Cytometry
Classification Monoclonal
Clone 645608
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q62230
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin, Siglec1
Gene Symbols SIGLEC1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Siglec-1/CD169 Thr20-Leu1639 (predicted) Accession # Q62230
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6614
Test Specificity Detects mouse Siglec-1/CD169 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Siglec-1, -5, -6, -7, -8, -9, -10, -11, -14, recombinant mouse Siglec-2, -3, -E, -F, -G, or -H is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 °C to 8 °C as supplied.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.