Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Siglec-2/CD22 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Siglec-2/CD22 |
---|---|
Clone | 308501 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Description
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Siglec-2/CD22 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
Cd22 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
308501 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
933 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-2/CD22 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Siglec-2/CD22 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -6, -7, -9, -10, -11, or recombinant mouse Siglec-F is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title