Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$208.00 - $2020.00
Specifications
Antigen | Siglec-F |
---|---|
Clone | 238047 |
Concentration | LYOPH |
Dilution | Neutralization 0.1-0.5 ug/mL |
Applications | Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17061SP
![]() |
R&D Systems
MAB17061SP |
25 μg |
Each for $208.00
|
|
|||||
MAB17061100
![]() |
R&D Systems
MAB17061100 |
100 μg |
Each for $582.50
|
|
|||||
MAB17061500
![]() |
R&D Systems
MAB17061500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Siglec-F Monoclonal specifically detects Siglec-F in Mouse samples. It is validated for Neutralization.Specifications
Siglec-F | |
LYOPH | |
Neutralization | |
Unconjugated | |
Rat | |
Mouse | |
Q920G3 | |
27181 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Siglec-F in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Siglec-2 or -3, or recombinant human Siglec-5, -7, -9, -10, or -11 is observed. |
238047 | |
Neutralization 0.1-0.5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
Siglecf | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title