Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SIGNR1/CD209b Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $526.50
Specifications
Antigen | SIGNR1/CD209b |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Adhesion Blockade 10 ug/mL |
Applications | Western Blot, Adhesion Blockade |
Classification | Polyclonal |
Description
SIGNR1/CD209b Polyclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blot, Adhesion Blockade.Specifications
SIGNR1/CD209b | |
Western Blot 0.1 ug/mL, Adhesion Blockade 10 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
Cd209b | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Adhesion Blockade | |
Unconjugated | |
RUO | |
Q8CJ91 | |
170786 | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1 Ser76-Gly325 Accession # Q8CJ91 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse SIGNR1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) SIGNR3 and rmSIGNR4 is observed, and approximately 10% cross-reactivity with recombinant human DC-SIGNR and rmDC-SIGN is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title