Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SIGNR1/CD209b Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1836
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Polyclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blot.Specifications
SIGNR1/CD209b | |
Polyclonal | |
Biotin | |
Q8CJ91 | |
Cd209b | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1/CD209b Ser76-Gly325 Accession # Q8CJ91 | |
50 μg | |
Primary | |
Detects mouse SIGNR1/CD209b in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse SIGNR4 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
Goat | |
Affinity Purified | |
RUO | |
170786 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction