Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SIGNR1/CD209b PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1836P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1836P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1836P Supplier R&D Systems Supplier No. FAB1836P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SIGNR1/CD209b Polyclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen SIGNR1/CD209b
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q8CJ91
Gene Alias CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7
Gene Symbols Cd209b
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1 Ser76-Gly325 Accession # Q8CJ91
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170786
Test Specificity Detects mouse SIGNR1 in direct ELISAs and Western blots. In Western blots, this antibody shows approximately 5% cross-reactivity with recombinant mouse SIGNR4.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.