Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SIGNR4/CD209e Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1528
This item is not returnable.
View return policy
Description
SIGNR4/CD209e Monoclonal specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot.Specifications
SIGNR4/CD209e | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q91ZW7 | |
Cd209e | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR4/CD209e Ser41-Trp208 Accession # Q91ZW7 | |
500 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
210006 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD209e, mSIGNR4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects mouse SIGNR4/CD209e in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SIGNR1 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction