Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SIGNR4/CD209e Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1528
This item is not returnable.
View return policy
Description
SIGNR4/CD209e Polyclonal specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot.Specifications
SIGNR4/CD209e | |
Polyclonal | |
Biotin | |
Q91ZW7 | |
Cd209e | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR4/CD209e Ser41-Trp208 Accession # Q91ZW7 | |
50 μg | |
Primary | |
Detects mouse SIGNR4/CD209e in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse SIGNR1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD209e, mSIGNR4 | |
Goat | |
Affinity Purified | |
RUO | |
170780 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction