Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SLAM/CD150 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB4330SP
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
SLAM/CD150 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q9QUM4 | |
SLAMF1 | |
Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150 Thr25-Pro242 Accession # Q9QUM4 | |
25 μg | |
Primary | |
Detects mouse SLAM/CD150. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF, ELISA | |
459911 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6504 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction