Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SLAM/CD150 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4330 Supplier R&D Systems Supplier No. AF4330
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SLAM/CD150 Polyclonal specifically detects SLAM/CD150 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen SLAM/CD150
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q9QUM4
Gene Alias CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1
Gene Symbols SLAMF1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150 Thr25-Pro242 Accession # Q9QUM4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6504
Test Specificity Detects mouse SLAM/CD150 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SLAM is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.