Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SLAM/CD150 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4330
This item is not returnable.
View return policy
Description
SLAM/CD150 Polyclonal specifically detects SLAM/CD150 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
SLAM/CD150 | |
Polyclonal | |
Biotin | |
Q9QUM4 | |
SLAMF1 | |
Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150 Thr25-Pro242 Accession # Q9QUM4 | |
50 μg | |
Primary | |
Detects mouse SLAM/CD150 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human SLAM, recombinant mouse (rm) 2B4/Fc, rmCD48, and rmCD84 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1 | |
Sheep | |
Affinity Purified | |
RUO | |
6504 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction