Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SLPI Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$227.50 - $561.50
Specifications
Antigen | SLPI |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
SLPI Polyclonal specifically detects SLPI in Mouse samples. It is validated for Western Blot.Specifications
SLPI | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
6590 | |
Mouse myeloma cell line NS0-derived mouse SLPI | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse SLPI in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human SLPI is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
SLPI | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title