Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Soggy-1/DkkL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1508
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1508 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1508 Supplier R&D Systems Supplier No. MAB1508
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Soggy-1/DkkL1 Monoclonal specifically detects Soggy-1/DkkL1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Soggy-1/DkkL1
Applications Western Blot
Classification Monoclonal
Clone 192425
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAF02679
Gene Alias Cancer/testis antigen 34, CT34dickkopf-like 1 (soggy), dickkopf-like 1, DkkL1, Protein soggy-1, SGY, SGY1, SGY-1dickkopf-like protein 1, SGY1soggy, soggy-1, Soggy1
Gene Symbols DKKL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Soggy-1/DkkL1 Leu21-Leu230 Accession # AAF02679
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27120
Test Specificity Detects mouse Soggy-1/DkkL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Soggy-1/DkkL1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.