Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Sonic Hedgehog/Shh C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB445
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB445 500 μg
MAB445SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB445 Supplier R&D Systems Supplier No. MAB445
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Sonic Hedgehog/Shh
Applications Western Blot
Classification Monoclonal
Clone 55626
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q62226.1
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gene Symbols SHH
Host Species Rat
Immunogen E. coli-derived recombinant mouse Sonic Hedgehog Cys199-Ser437 Accession # Q62226.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6469
Test Specificity Detects recombinant mouse Shh C-terminal peptide in direct ELISAs and Western blots. Does not cross-react in direct ELISAs with rmDhh C- or N-terminal peptides, rmIhh C- or N-terminal peptides, rhShh N-terminal peptide, or rmShh N-terminal peptide.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.