Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Sonic Hedgehog/Shh C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF445
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF445 Supplier R&D Systems Supplier No. AF445
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 8 publications

Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Sonic Hedgehog/Shh
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q62226
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gene Symbols SHH
Host Species Goat
Immunogen E. coli-derived recombinant mouse Sonic Hedgehog Cys25-Ser437 with a Lys122Arg substitution Accession # Q62226
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6469
Test Specificity Detects mouse Shh C-terminal peptide in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant mouse (rm) Dhh C-terminal peptide is observed and less than 1% cross-reactivity with a 6X histidine-tagged rmShh N-terminal peptide (amino acids 25-198) is observed. Detects human and mouse Sonic Hedgehog protein in Western blots. Shh C-terminal peptide specific IgG was purified by first passing the goat sera over a mouse Shh N-terminal peptide affinity column. The unbound fraction from the mouse Shh N-terminal peptide affinity column was subsequently purified using a mouse Shh C-terminal peptide affinity column.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.