Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SorCS3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30671SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30671SP 25 μg
MAB30671 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB30671SP Supplier R&D Systems Supplier No. MAB30671SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

SorCS3 Monoclonal specifically detects SorCS3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SorCS3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 339615
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8VI51
Gene Alias 6330404A12Rik, KIAA1059SORCS, sortilin-related VPS10 domain containing receptor 3, VPS10 domain receptor protein SORCS 3 (SORCS3), VPS10 domain-containing receptor SorCS3
Gene Symbols SORCS3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Glu34-Ser1122 Accession # Q8VI51
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22986
Test Specificity Detects human and mouse SorCS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SorCS1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.