Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SOST/Sclerostin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1589SP
This item is not returnable.
View return policy
Description
SOST/Sclerostin Polyclonal specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SOST/Sclerostin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
sclerostin, VBCHsclerosteosis | |
Goat | |
Affinity Purified | |
RUO | |
50964 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_077769 | |
SOST | |
Mouse myeloma cell line NS0-derived recombinant mouse SOST/Sclerostin Gln24-Tyr211 Accession # NP_077769 | |
25 μg | |
Primary | |
Detects mouse SOST/Sclerostin in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human SOST is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction