Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SPARC-like 1/SPARCL1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | SPARC-like 1/SPARCL1 |
---|---|
Clone | 348905 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Mouse samples. It is validated for Western Blot.Specifications
SPARC-like 1/SPARCL1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P70663 | |
8404 | |
Mouse myeloma cell line NS0-derived recombinant mouse SPARC-like 1/SPARCL1 Ile17-Phe650 Accession # P70663 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse SPARC-like 1/SPARCL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SPARC or recombinant human SPARC-like 1 is observed. |
348905 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC like 1, SPARCL1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
SPARCL1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title