Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SPARC-like 1/SPARCL1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$214.00 - $585.00
Specifications
Antigen | SPARC-like 1/SPARCL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
SPARC-like 1/SPARCL1 Polyclonal specifically detects SPARC-like 1/SPARCL1 in Mouse samples. It is validated for Western Blot.Specifications
SPARC-like 1/SPARCL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC like 1, SPARCL1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
SPARCL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P70663 | |
8404 | |
Mouse myeloma cell line NS0-derived recombinant mouse SPARC-like 1/SPARCL1 (R&D Systems, Catalog # 4547-SL) Ile17-Phe650 Accession # P70663 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse SPARC-like 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human SPARC-like 1 is observed and less than 1% cross-reactivity with recombinant mouse SPARC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title