Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Spi-B Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF7204SP
This item is not returnable.
View return policy
Description
Spi-B Polyclonal specifically detects Spi-B in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Spi-B | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SpiB, SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
Sheep | |
Affinity Purified | |
RUO | |
6689 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
O35906 | |
SPIB | |
E. coli-derived recombinant mouse Spi-B Tyr18-Glu167 (Tyr110Phe) Accession # O35906 | |
25 μg | |
Primary | |
Detects mouse Spi-B in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human Spi-B is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction