Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Spi-B Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF7204
This item is not returnable.
View return policy
Description
Spi-B Polyclonal specifically detects Spi-B in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Spi-B | |
Polyclonal | |
Unconjugated | |
O35906 | |
SPIB | |
E. coli-derived recombinant mouse Spi-B Tyr18-Glu167 (Tyr110Phe) Accession # O35906 | |
100 μg | |
Primary | |
Detects mouse Spi-B in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human Spi-B is observed. | |
Mouse | |
IgG |
Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
SpiB, SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
Sheep | |
Affinity Purified | |
RUO | |
6689 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction