Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SR-AI/MSR Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1797R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1797R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1797R100 Supplier R&D Systems Supplier No. FAB1797R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen SR-AI/MSR
Applications Flow Cytometry
Classification Monoclonal
Clone 268318
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAA39747
Gene Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI
Gene Symbols MSR1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SR-AI/MSR Trp79-Ser454 Accession # AAA39747
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4481
Test Specificity Detects mouse SR-AI/MSR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SR-AI is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.