Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ST2/IL-33R APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB10041A025
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB10041A025 25 μg
FB10041A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FB10041A025 Supplier R&D Systems Supplier No. FAB10041A025UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ST2/IL-33R
Applications Flow Cytometry
Classification Monoclonal
Clone 245707
Conjugate APC
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P14719
Gene Alias DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Gene Symbols Il1rl1
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ST2/IL-33 R, Ser27-Arg332, Accession No. P14719
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 9173.0
Test Specificity Detects mouse ST2/IL-33 R in ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.