Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Stella/Dppa3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2566
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2566 100 μg
AF2566SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2566 Supplier R&D Systems Supplier No. AF2566
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Stella/Dppa3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 μg/mL, Immunohistochemistry 1-15 μg/mL
Gene Accession No. Q8QZY3
Gene Alias developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
Gene Symbols DPPA3
Host Species Goat
Immunogen E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150 Accession # Q8QZY3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359787
Test Specificity Detects mouse Stella/Dppa3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Stella is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.