Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Syndecan-1/CD138 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2966R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2966R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2966R100 Supplier R&D Systems Supplier No. FAB2966R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Syndecan-1/CD138
Applications Flow Cytometry
Classification Monoclonal
Clone 300506
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P18828
Gene Alias CD138, CD138 antigen, SDC, SDC1, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1, Syndecan1
Gene Symbols SDC1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-1/CD138 isoform 1 Gln18-Glu252 Accession # P18828
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6382
Test Specificity Detects mouse Syndecan-1/CD138 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Syndecan-1, recombinant mouse (rm) Syndecan-3 or rmSyndecan-4 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.