Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Tau ELISA Kit (Colorimetric)
SDP

Catalog No. NB016819 Shop All R&D Systems Products
Change view
Click to view available options
Interassay CV:
0.0472
0.0527
Intraassay CV:
0.0466
0.0551
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Interassay CV Intraassay CV
NB016819 0.0527 0.0466
NB016722 0.0472 0.0551
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016819 Supplier Novus Biologicals™ Supplier No. NBP281260
Add to Cart
Add to Cart

This kit recognizes Mouse Tau in samples.

No significant cross-reactivity or interference between Mouse Tau and analogues was observed.

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse pMAPT/pTAU in samples. No significant cross-reactivity or interference between Mouse pMAPT/pTAU and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 4137
Gene Symbol MAPT
Host Species Mouse
Interassay CV 0.0527
Intraassay CV 0.0466
Quantity 1 Kit
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Tau
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.