Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TCF-3/E2A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7650SP
This item is not returnable.
View return policy
Description
TCF-3/E2A Polyclonal specifically detects TCF-3/E2A in Mouse samples. It is validated for Western Blot.Specifications
TCF-3/E2A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1, ITF1Transcription factor ITF-1, Kappa-E2-binding factor, MGC129647, MGC129648, Pan2, Tcfe2a, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
Sheep | |
Affinity Purified | |
RUO | |
6929 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P15806 | |
TCF3 | |
E. coli-derived recombinant mouse TCF-3/E2A Asn33-Arg159 Accession # P15806 | |
25 μg | |
Primary | |
Detects mouse TCF-3/E2A in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction