Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TCR gamma/delta Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7297U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7297U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7297U100 Supplier R&D Systems Supplier No. FAB7297U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hamster Monoclonal Antibody

TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry
Classification Monoclonal
Clone GL-3
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias delta polypeptide, T cell receptor gamma locus, T3D, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TiT3, TRG
Gene Symbols Cd3d
Host Species Hamster
Immunogen Mouse intraepithelial lymphocytes
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6965
Test Specificity Detects mouse TCR gamma/delta .
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.