Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TCR gamma/delta Alexa Fluor 405-conjugated Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems FAB7297V100UG
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR gamma/delta | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
delta polypeptide, T cell receptor gamma locus, T3D, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TiT3, TRG | |
Hamster | |
100 μg | |
Primary | |
Detects mouse TCR gamma/delta . | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
GL-3 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Cd3d | |
Mouse intraepithelial lymphocytes | |
RUO | |
6965 | |
Mouse | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction