Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TCR gamma/delta Antibody, R&D Systems™

Hamster Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB7297SP
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry, CyTOF-reported.Specifications
| TCR gamma/delta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cd3d | |
| Mouse intraepithelial lymphocytes | |
| 25 μg | |
| Primary | |
| Detects mouse TCR gamma/delta . | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| GL-3 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
| delta polypeptide, T cell receptor gamma locus, T3D, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TiT3, TRG | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6965 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction