Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TCR gamma/delta Biotinylated Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems FAB7297B100
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR gamma/delta | |
Monoclonal | |
Biotin | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6965 | |
Mouse | |
Purified |
Flow Cytometry | |
GL-3 | |
Flow Cytometry 10 uL/10^6 cells | |
delta polypeptide, T cell receptor gamma locus, T3D, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TiT3, TRG | |
Mouse intraepithelial lymphocytes | |
100 Tests | |
Primary | |
Detects mouse TCR gamma/delta in flow cytometry. | |
Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction