Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Tenascin C ELISA Kit (Colorimetric)
SDP

Catalog No. NB001553
Click to view available options
Quantity:
1 Kit

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse TNC in samples. No significant cross-reactivity or interference between Mouse TNC and analogues was observed
For Use With (Application) ELISA
Gene ID (Entrez) 3371
Gene Symbol TNC
Host Species Mouse
Quantity 1 Kit
Research Discipline Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Tenascin C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.