Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TER-119 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1125G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1125G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1125G100 Supplier R&D Systems Supplier No. FAB1125G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TER-119 Monoclonal specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TER-119
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias Ly76, Ly-76, TER119
Gene Symbols Ly76
Host Species Rat
Immunogen C57BL/6 mouse day-14 fetal liver cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104231
Test Specificity Detects mouse TER-119 in Western blots. This antibody has been shown to react with cells of the erythroid lineage in embryonic yolk sac, fetal liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs (Ikuta, K. et al. (1990) Cell 62:863). TER-119 Antigen is a molecule associated with cell-surface glycophorin A (Kina, T. et al. (2000) Br. J. Haematol. 109:280). The antibody has been shown to react with erythroid cells at differentiation stages from early proerythroblast to mature erythrocyte (Kina, T. et al. (2000) Br. J. Haematol. 109:280).
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.