Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Testican 3/SPOCK3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Testican 3/SPOCK3 |
---|---|
Clone | 330404 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB23461SP
![]() |
R&D Systems
MAB23461SP |
25 μg |
Each for $200.50
|
|
|||||
MAB23461
![]() |
R&D Systems
MAB23461 |
100 μg |
Each for $491.00
|
|
|||||
Description
Testican 3/SPOCK3 Monoclonal specifically detects Testican 3/SPOCK3 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Testican 3/SPOCK3 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
Q8BKV0 | |
50859 | |
Mouse myeloma cell line NS0-derived recombinant mouse Testican 3/SPOCK3 Val26-Ile436 Accession # Q8BKV0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Testican 3/SPOCK3 in direct ELISAs and Western blots. In Western blots, approximately 30% cross-reactivity with recombinant human (rh) Testican 1 and rhTestican 3 is observed. |
330404 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, SPOCK3, TES-3, testican 3, TICN3 | |
SPOCK3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title