Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TEX19.1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6645
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6645 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6645 Supplier R&D Systems Supplier No. MAB6645
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TEX19.1 Monoclonal specifically detects TEX19.1 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen TEX19.1
Applications Immunocytochemistry
Classification Monoclonal
Clone 678803
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99MV2
Gene Alias TEX19, Tex19a
Gene Symbols Tex19.1
Host Species Rat
Immunogen E. coli-derived recombinant mouse TEX19.1 Arg189-Pro351 Accession # Q99MV2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400629
Test Specificity Detects mouse TEX19.1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human TEX19, recombinant mouse (rm) TEX19.2, or rmTEX11 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.