Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TFPI Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2975SP
This item is not returnable.
View return policy
Description
TFPI Monoclonal specifically detects TFPI in Mouse samples. It is validated for Western Blot.Specifications
TFPI | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O54819 | |
TFPI | |
Mouse myeloma cell line NS0-derived recombinant mouse TFPI Leu29-Lys289 Accession # O54819 | |
25 μg | |
Primary | |
Detects mouse TFPI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with recombinant human (rh) TFPI or rmTFPI-2. | |
Mouse | |
Purified |
Western Blot | |
372311 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7035 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction