Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TGF-beta RI/ALK-5 APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Specifications
Antigen | TGF-beta RI/ALK-5 |
---|---|
Clone | 141231 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5871A025
|
R&D Systems
FAB5871A025 |
25 Tests | N/A | N/A | N/A | ||||
FAB5871A100
|
R&D Systems
FAB5871A100 |
100 Tests | N/A | N/A | N/A | ||||
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Flow Cytometry.Specifications
TGF-beta RI/ALK-5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
TGFBR1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detect mouse TGF-beta RI/ALK-5 in flow cytometry. Although this clone detects human and mouse TGF-beta RI/ALK-5 in Western blots, positive results have not been observed on human TGF-beta RI/ALK-5 in flow cytometry. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with rrMIS RII, rhTGF-beta RII, rhTGF-beta RIIb, or rhTGF-beta RIII. |
141231 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
BAA05023 | |
7046 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title