Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Thrombopoietin R/Tpo R Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1317
This item is not returnable.
View return policy
Description
Thrombopoietin R/Tpo R Polyclonal specifically detects Thrombopoietin R/Tpo R in Mouse samples. It is validated for Western Blot.Specifications
Thrombopoietin R/Tpo R | |
Polyclonal | |
Biotin | |
CAA52031 | |
MPL | |
Mouse myeloma cell line NS0-derived recombinant mouse Thrombopoietin R/Tpo R Ser18-Trp483 Accession # CAA52031 | |
50 μg | |
Primary | |
Detects mouse Thrombopoietin R/Tpo R in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human Tpo R and recombinant mouse Epo R is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD110, CD110 antigen, C-MPL, MPL, MPLV, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TpoR, TPO-R, TPORMPLV | |
Goat | |
Affinity Purified | |
RUO | |
4352 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction