Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Thrombopoietin/Tpo Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF488
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Polyclonal specifically detects Thrombopoietin/THPO in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Thrombopoietin/Tpo | |
Polyclonal | |
Biotin | |
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
Goat | |
Affinity Purified | |
RUO | |
7066 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
THPO | |
Mouse myeloma cell line NS0-derived recombinant mouse Thrombopoietin/Tpo and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Thrombopoietin/Tpo | |
50 μg | |
Primary | |
Detects mouse Thrombopoietin/Tpo in ELISAs and Western blots. When used in combination with the monoclonal capture antibody (Catalog # MAB488) in sandwich immunoassays, less than 1% cross-reactivity with NS0-derived recombinant human Tpo is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction