Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Thrombospondin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7860
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7860 Supplier R&D Systems Supplier No. MAB7860
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Thrombospondin-4 Monoclonal specifically detects Thrombospondin-4 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Thrombospondin-4
Applications Immunohistochemistry
Classification Monoclonal
Clone 893655
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Gene Accession No. Q9Z1T2
Gene Alias THBS4, thrombospondin 4, thrombospondin-4, Thrombospondin4, TSP4, TSP-4
Gene Symbols THBS4
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Thrombospondin-4 Gln27-Asn963 (predicted) Accession # Q9Z1T2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7060
Test Specificity Detects mouse Thrombospondin-4 in ELISA.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.